Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl borate, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
688-74-4
C12H27BO3
230.15
LGQXXHMEBUOXRP-UHFFFAOYSA-N
n-butyl borate,tri-n-butyl borate,tributoxyborane,butyl borate,tributoxyboron,boron tributoxide,borester 2,tri-n-butoxyborane,tris butoxy borane,borane, tributoxy
12712
tributyl borate
B(OCCCC)(OCCCC)OCCCC
688-74-4
C12H27BO3
230.15
LGQXXHMEBUOXRP-UHFFFAOYSA-N
n-butyl borate,tri-n-butyl borate,tributoxyborane,butyl borate,tributoxyboron,boron tributoxide,borester 2,tri-n-butoxyborane,tris butoxy borane,borane, tributoxy
12712
tributyl borate
B(OCCCC)(OCCCC)OCCCC
Quantity 2.5L
Density 0.8530g/mL
Specific Gravity 0.853
Refractive Index 1.4080 to 1.4100
Color Colorless
Flash Point 93°C
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point -70.0°C
Boiling Point 230.0°C to 235.0°C
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH3CH2CH2CH2O)3B
Beilstein 01,II,398
Fieser 13,326
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with most common organic liquids
Formula Weight 230.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Tributyl borate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 211-706-5