Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
100mL
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
102-82-9
C12H27N
185.35
MFCD00009431
IMFACGCPASFAPR-UHFFFAOYSA-N
tributylamine,tri-n-butylamine,1-butanamine, n,n-dibutyl,n,n-dibutyl-1-butanamine,tributilamina,tris-n-butylamine,amine, tributyl,tris n-butylamine,tributilamina romanian,unii-c3tzb2w0r7
7622
CHEBI:38905
N,N-dibutylbutan-1-amine
CCCCN(CCCC)CCCC
102-82-9
C12H27N
185.35
MFCD00009431
IMFACGCPASFAPR-UHFFFAOYSA-N
tributylamine,tri-n-butylamine,1-butanamine, n,n-dibutyl,n,n-dibutyl-1-butanamine,tributilamina,tris-n-butylamine,amine, tributyl,tris n-butylamine,tributilamina romanian,unii-c3tzb2w0r7
7622
CHEBI:38905
N,N-dibutylbutan-1-amine
CCCCN(CCCC)CCCC
Melting Point -70.0°C
Quantity 250mL
Specific Gravity 0.778
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4273 to 1.4293
Density 0.7780g/mL
Color Undesignated
Boiling Point 215.0°C
Flash Point 75°C
Viscosity 2.4 mPa.s (20°C)
Water 0.2% max.
Assay Percent Range 99%
Linear Formula [CH3NCH2)3]3N
Beilstein 04,157
Fieser 01,1189; 06,601
Merck Index 15,978
Solubility Information Solubility in water: 0.08g/L water (20°C). Other solubilities: soluble in most organic solvents,very soluble in alcohol and ether
Formula Weight 185.35
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Tributylamine
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Fatal if inhaled.
Toxic in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger