Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributylphosphine oxide, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
814-29-9
C12H27OP
218.321
MFCD00002082
MNZAKDODWSQONA-UHFFFAOYSA-N
tributylphosphine oxide,tri-n-butylphosphine oxide,butyphos,phosphine oxide, tributyl,trisbutylphosphine oxide,tributylfosfinoxid,tributylfosfinoxid czech,unii-w58cdf8s4h,tributylphosphane oxide,w58cdf8s4h
13138
1-dibutylphosphorylbutane
CCCCP(=O)(CCCC)CCCC
814-29-9
C12H27OP
218.321
MFCD00002082
MNZAKDODWSQONA-UHFFFAOYSA-N
tributylphosphine oxide,tri-n-butylphosphine oxide,butyphos,phosphine oxide, tributyl,trisbutylphosphine oxide,tributylfosfinoxid,tributylfosfinoxid czech,unii-w58cdf8s4h,tributylphosphane oxide,w58cdf8s4h
13138
1-dibutylphosphorylbutane
CCCCP(=O)(CCCC)CCCC
Color White
Quantity 10g
Melting Point 64°C to 69°C
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula [CH3(CH2)3]3P(O)
Beilstein 04, II, 974
Fieser 01,1192; 15,333
Formula Weight 218.32
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Tributylphosphine oxide, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-394-3

RTECSNumber : SZ1575000

TSCA : TSCA