Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyltin trifluoromethanesulfonate, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
68725-14-4
C13H27F3O3SSn
439.13
MFCD00043324
68725-14-4
C13H27F3O3SSn
439.13
MFCD00043324
Packaging Glass bottle
Boiling Point 56°C to 60°C (25.0mmHg)
Melting Point 31°C
Flash Point >110°C
Infrared Spectrum Authentic
Color White to Beige
Quantity 1g
Assay Percent Range 96%
Formula Weight 439.13
Physical Form Crystals and Chunks
Percent Purity 96%
Chemical Name or Material Tributyltin trifluoromethanesulfonate
Show More Show Less

GHS H Statement
Causes damage to organs through prolonged or repeated exposure. Causes serious eye damage. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. Toxic if swallowed. Harmful in contact with skin. Very toxic to aquatic life.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

GHS Signal Word: Danger