Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyrin, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
250mL
1L
Packaging:
Glass bottle
60-01-5
C15H26O6
302.36
MFCD00009392
UYXTWWCETRIEDR-UHFFFAOYSA-N
tributyrin,glyceryl tributyrate,glycerol tributyrate,tributin,butyrin,butyryl triglyceride,propane-1,2,3-triyl tributyrate,glycerol tributanoate,glyceroltributyrin,tri-n-butyrin
6050
CHEBI:35020
2,3-di(butanoyloxy)propyl butanoate
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC
60-01-5
C15H26O6
302.36
MFCD00009392
UYXTWWCETRIEDR-UHFFFAOYSA-N
tributyrin,glyceryl tributyrate,glycerol tributyrate,tributin,butyrin,butyryl triglyceride,propane-1,2,3-triyl tributyrate,glycerol tributanoate,glyceroltributyrin,tri-n-butyrin
6050
CHEBI:35020
2,3-di(butanoyloxy)propyl butanoate
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC
Refractive Index 1.4340 to 1.437
Quantity 1L
Appearance Liquid
Packaging Glass bottle
Infrared Spectrum Conforms
Color Clear colorless to light yellow
Flash Point 173°C
Identification pass test
Specific Gravity 1.0335
Boiling Point 305.0°C to 310.0°C
Density 1.0335g/mL
Melting Point -75.0°C
Assay Percent Range 97%
Linear Formula (CH3CH2CH2CO2CH2)2CH(O2CCH2CH2CH3)
Beilstein 02, 273
Merck Index 15, 9785
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 302.36
Physical Form Liquid
Percent Purity 0.97
Chemical Name or Material Tributyrin, 98%
Show More Show Less

EINECSNumber : 200-451-5

RTECSNumber : ET7350000

TSCA : TSCA