Alfa Aesar™ Tricarbonylnitrosylcobalt

Manufacturer: Alfa Aesar™ 2315106

Quantity 5g

 View more versions of this product

Catalog No. AA2315106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14096-82-3
    Linear Formula Co(NO)(CO)3
    Formula Weight 172.97
    MDL Number MFCD00016014
    UN Number UN1992
    Synonym Cobalt tricarbonyl nitrosyl
    Chemical Name or Material Tricarbonylnitrosylcobalt

    Specifications

    Formula Weight 172.97
    Health Hazard 3 GHS P Statement
    P210-P280h-P304+P340-P310-P302+P352
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Color Dark red
    Odor Unpleasant
    Physical Form Liquid
    MDL Number MFCD00016014
    Density 1.47g/mL
    UN Number UN1992
    Boiling Point 48.6°C (55°C (dec.))
    Sensitivity Air Sensitive
    Synonym Cobalt tricarbonyl nitrosyl
    Melting Point −1°C
    Health Hazard 1 Danger
    Quantity 5g
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    Linear Formula Co(NO)(CO)3
    CAS 14096-82-3
    Health Hazard 2 GHS H Statement
    H225-H301-H330-H351-H317
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Fatal if inhaled.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Chemical Name or Material Tricarbonylnitrosylcobalt

    GHS H Statement
    H225-H301-H330-H351-H317
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Fatal if inhaled.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P280h-P304+P340-P310-P302+P352
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit