Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tricarbonylnitrosylcobalt

Manufacturer:  Alfa Aesar™ 2315103

Quantity 1g

 View more versions of this product

Catalog No. AA2315103


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14096-82-3
172.97
UN1992
Tricarbonylnitrosylcobalt
Co(NO)(CO)3
MFCD00016014
Cobalt tricarbonyl nitrosyl

Specifications

14096-82-3
Co(NO)(CO)3
MFCD00016014
UN1992
−1°C
48.6°C (55°C (dec.))
Unpleasant
Tricarbonylnitrosylcobalt
Liquid
172.97
1.47g/mL
Cobalt tricarbonyl nitrosyl
Dark red
1g
Air Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H301-H330-H351-H317
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P210-P280h-P304+P340-P310-P302+P352
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit