Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trichloro(1H,1H,2H,2H-heptadecafluorodecyl)silane 96.0+%, TCI America™
Click to view available options
Quantity:
5g
25g
Specifications
Quantity | 5g |
Color | Colorless |
Boiling Point | 118°C |
UN Number | 2987 |
Formula Weight | 581.55 |
Physical Form | Clear Liquid at 20°C |
Percent Purity | ≥96.0% (GC) |
Chemical Name or Material | Trichloro(1H,1H,2H,2H-heptadecafluorodecyl)silane |
Safety and Handling
EINECSNumber : (2)-2046
TSCA : Yes