Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, 3% (w/v) Aqueous Solution, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
CAS 76-03-9, 7732-18-5
Percent Purity 3% ± 0.3%
Grade Chemical Solution
Quantity 500 mL
Packaging Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.