Learn More
Trichloroacetic acid, 99%, Thermo Scientific Chemicals
Description
Protein precipitation reagent Trichloroacetic acid is used as a precipitating agent in biochemistry for precipitation of proteins, DNA and RNA. It is an active ingredient used for cosmetic treatments like chemical peels, tattoo removal and the treatment of warts including genital warts. It is also used to determine protein concentration and a decalcifier and fixative in microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Protein precipitation reagent Trichloroacetic acid is used as a precipitating agent in biochemistry for precipitation of proteins, DNA and RNA. It is an active ingredient used for cosmetic treatments like chemical peels, tattoo removal and the treatment of warts including genital warts. It is also used to determine protein concentration and a decalcifier and fixative in microscopy.
Solubility
Soluble in water, alcohol, ethyl ether, benzene, acetone, o-xylene and carbon tetrachloride.
Notes
Moisture sensitive. Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents, strong bases and amines.
Specifications
pH | <1 |
Melting Point | 54°C to 58°C |
Boiling Point | 196°C |
UN Number | UN1839 |
Beilstein | 970119 |
Merck Index | 14,9627 |
Solubility Information | Soluble in water,alcohol,ethyl ether,benzene,acetone,o-xylene and carbon tetrachloride. |
Formula Weight | 163.39 |
Percent Purity | 99% |
Density | 1.629 |
Show More |
Safety and Handling
Hazard Category | H314-H335-H351 |
Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
Precautionary Statement | P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TRICHLOROACETIC ACID |
EINECSNumber | 200-927-2 |
RTECSNumber | AJ7875000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.