Alfa Aesar™ Trichloroacetic acid, 99%

Manufacturer: Alfa Aesar™ A1115636

Quantity 500g

 View more versions of this product

Catalog No. AAA1115636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76-03-9
    Molecular Formula Cl3CCO2H
    Formula Weight 163.39
    MDL Number MFCD00004177
    UN Number UN1839
    Beilstein 970119
    Merck Index 14,9627
    Synonym TCA
    Chemical Name or Material Trichloroacetic acid

    Specifications

    Formula Weight 163.39
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 196°C
    Quantity 500g
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TRICHLOROACETIC ACID
    TSCA TSCA
    Melting Point 54° to 58°C
    Beilstein 970119
    Synonym TCA
    pH <1
    Molecular Formula Cl3CCO2H
    MDL Number MFCD00004177
    Merck Index 14,9627
    Odor Acrid
    CAS 76-03-9
    UN Number UN1839
    Health Hazard 1 Danger
    RTECS Number AJ7875000
    Chemical Name or Material Trichloroacetic acid
    Assay 99%

    Description

    Applications
    Protein precipitation reagent Trichloroacetic acid is used as a precipitating agent in biochemistry for precipitation of proteins, DNA and RNA. It is an active ingredient used for cosmetic treatments like chemical peels, tattoo removal and the treatment of warts including genital warts. It is also used to determine protein concentration and a decalcifier and fixative in microscopy.

    Solubility
    Soluble in water, alcohol, ethyl ether, benzene, acetone, o-xylene and carbon tetrachloride.

    Notes
    Moisture sensitive. Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents, strong bases and amines.

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit