Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic acid, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
2.5kg
10kg
Packaging:
Plastic Drum
Plastic bottle
76-03-9
C2HCl3O2
163.39
MFCD00004177
YNJBWRMUSHSURL-UHFFFAOYSA-N
trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloracetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer
6421
CHEBI:30956
2,2,2-trichloroacetic acid
C(=O)(C(Cl)(Cl)Cl)O
76-03-9
C2HCl3O2
163.39
MFCD00004177
YNJBWRMUSHSURL-UHFFFAOYSA-N
trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloracetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer
6421
CHEBI:30956
2,2,2-trichloroacetic acid
C(=O)(C(Cl)(Cl)Cl)O
Color White
Packaging Plastic bottle
Density 1.6200g/mL
Specific Gravity 1.62
Water 0.5% max. (K.F.)
Boiling Point 196.0°C
Quantity 1kg
Melting Point 52.0°C to 58.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula CCl3CO2H
Beilstein 02,206
Fieser 01,1194; 02,425; 04,520; 15,168
Merck Index 15,9792
Solubility Information Solubility in water: 120g/100 mL (20°C). Other solubilities: soluble in alcohol and ether,carbon tetrachloride
Formula Weight 163.39
Physical Form Flakes, Crystals, or Crystalline Powder
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Trichloroacetic acid, Extra Pure
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger