Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, Crystal, 95-100.5%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
76-03-9
C2HCl3O2
163.38
YNJBWRMUSHSURL-UHFFFAOYSA-N
trichloroacetic acid
OC(=O)C(Cl)(Cl)Cl
76-03-9
C2HCl3O2
163.38
YNJBWRMUSHSURL-UHFFFAOYSA-N
trichloroacetic acid
OC(=O)C(Cl)(Cl)Cl

TR111, 76-03-9, C2HCl3O2

Spectrum™ Trichloroacetic Acid, Crystal , also known as TCA, is used in biochemistry to help the precipitation of macromolecules. It is also used in the cosmetic industry as a chemical peel and also as a wart remover.

CAS Max % 1
Percent Purity 95.0 to 100.5%
Grade Reagent
Ignition Residue 0.0005
Packaging Poly Pail
Sulfate 0.0008
Quantity 25 kg