Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
2.5kg
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

76-03-9
C2HCl3O2
163.378
YNJBWRMUSHSURL-UHFFFAOYSA-N
6421
CHEBI:30956

High quality chemicals for laboratory and specialized industrial use

CAS 76-03-9
Molecular Formula C2HCl3O2
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES C(=O)(C(Cl)(Cl)Cl)O
IUPAC Name 2,2,2-trichloroacetic acid
Molecular Weight (g/mol) 163.378
PubChem CID 6421
ChEBI CHEBI:30956
Formula Weight 163.39
Physical Form Crystals
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.60kg
Quantity 2.5kg
Chemical Name or Material Trichloroacetic Acid
Grade Baker Analyzed™ ACS Reagent
Show More Show Less