Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
76-03-9
C2HCl3O2
163.378
YNJBWRMUSHSURL-UHFFFAOYSA-N
trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloracetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer
6421
CHEBI:30956
2,2,2-trichloroacetic acid
C(=O)(C(Cl)(Cl)Cl)O
76-03-9
C2HCl3O2
163.378
YNJBWRMUSHSURL-UHFFFAOYSA-N
trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloracetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer
6421
CHEBI:30956
2,2,2-trichloroacetic acid
C(=O)(C(Cl)(Cl)Cl)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 163.39
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.60kg
Quantity 500g
Chemical Name or Material Trichloroacetic Acid