Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic acid, sodium salt, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
650-51-1
C2Cl3NaO2
185.36
MFCD00064198
SAQSTQBVENFSKT-UHFFFAOYSA-M
sodium trichloroacetate,tca-sodium,trichloroacetic acid sodium salt,antiperz,antyperz,tca sodium,acp grass killer,sodium tca,varitox,stca
23681045
CHEBI:81976
sodium;2,2,2-trichloroacetate
C(=O)(C(Cl)(Cl)Cl)[O-].[Na+]
650-51-1
C2Cl3NaO2
185.36
MFCD00064198
SAQSTQBVENFSKT-UHFFFAOYSA-M
sodium trichloroacetate,tca-sodium,trichloroacetic acid sodium salt,antiperz,antyperz,tca sodium,acp grass killer,sodium tca,varitox,stca
23681045
CHEBI:81976
sodium;2,2,2-trichloroacetate
C(=O)(C(Cl)(Cl)Cl)[O-].[Na+]
Assay Percent Range 97%
Linear Formula CCl3CO2Na
Fieser 01,1107; 02,388; 06,553; 07,341
Merck Index 15, 9792
Formula Weight 185.37
Physical Form Powder or Granular Powder
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Solubility in water: soluble (153 g/100mL at 20°C). Other solubilities: soluble in ethanol, acetone, diethyl ether
Melting Point >300.0°C
Color White
Quantity 2.5kg
Chemical Name or Material Trichloroacetic acid, sodium salt
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 211-479-2

RTECSNumber : AJ9100000

TSCA : TSCA