Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
545-06-2
C2Cl3N
144.39
DRUIESSIVFYOMK-UHFFFAOYSA-N
trichloroacetonitrile,acetonitrile, trichloro,tritox,cyanotrichloromethane,trichloromethyl cyanide,trichloroethanenitrile,trichlouracetonitril,trichlor-acetonitril,trichloromethylnitrile,nitrile trichloracetique
11011
CHEBI:82541
2,2,2-trichloroacetonitrile
C(#N)C(Cl)(Cl)Cl
545-06-2
C2Cl3N
144.39
DRUIESSIVFYOMK-UHFFFAOYSA-N
trichloroacetonitrile,acetonitrile, trichloro,tritox,cyanotrichloromethane,trichloromethyl cyanide,trichloroethanenitrile,trichlouracetonitril,trichlor-acetonitril,trichloromethylnitrile,nitrile trichloracetique
11011
CHEBI:82541
2,2,2-trichloroacetonitrile
C(#N)C(Cl)(Cl)Cl
Boiling Point 83.0°C to 84.0°C
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Melting Point -42.0°C
Infrared Spectrum Authentic
Quantity 2.5kg
Density 1.4400g/mL
Color Colorless to Yellow
Specific Gravity 1.44
Flash Point 195°C
Assay Percent Range 98%
Linear Formula CCl3CN
Beilstein 02,212
Fieser 01,1194; 05,686; 06,604; 07,381; 12,526; 13,32
Merck Index 15,9793
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,dimethyl sulfoxide,ethanol
Formula Weight 144.39
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Trichloroacetonitrile
Show More Show Less