Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetyl isocyanate, NMR grade, ACROS Organics™

Click to view available options
Quantity:
2g
10g
50g
Packaging:
Glass bottle
3019-71-4
C3Cl3NO2
188.39
MFCD00002033
GRNOZCCBOFGDCL-UHFFFAOYSA-N
trichloroacetyl isocyanate, acetyl isocyanate, trichloro, trichloroacetylisocyanate, isocyanic acid trichloroacetyl ester, unii-qxv1p5j6jl, qxv1p5j6jl, acetyl isocyanate, 2,2,2-trichloro, 2,2,2-trichloroethanecarbonyl isocyanate, trichloroethanecarbonyl isocyanate, trichloroacetyl isocyanate, nmr grade
76400
2,2,2-trichloroacetyl isocyanate
ClC(Cl)(Cl)C(=O)N=C=O
3019-71-4
C3Cl3NO2
188.39
MFCD00002033
GRNOZCCBOFGDCL-UHFFFAOYSA-N
trichloroacetyl isocyanate, acetyl isocyanate, trichloro, trichloroacetylisocyanate, isocyanic acid trichloroacetyl ester, unii-qxv1p5j6jl, qxv1p5j6jl, acetyl isocyanate, 2,2,2-trichloro, 2,2,2-trichloroethanecarbonyl isocyanate, trichloroethanecarbonyl isocyanate, trichloroacetyl isocyanate, nmr grade
76400
2,2,2-trichloroacetyl isocyanate
ClC(Cl)(Cl)C(=O)N=C=O
Packaging Glass bottle
Quantity 10g
Boiling Point 80.0°C to 85.0°C (20.0mmHg)
Infrared Spectrum Authentic
Refractive Index 1.4799 to 1.4819
Flash Point 65°C
Density 1.5900g/mL
Color Colorless to Yellow
Specific Gravity 1.59
Linear Formula CCl3CONCO
Fieser 02,425; 07,382
Solubility Information Solubility in water: decomposes
Formula Weight 188.4
Physical Form Liquid
Percent Purity ≥97%
Chemical Name or Material Trichloroacetyl isocyanate
Show More Show Less

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-165-7

TSCA : TSCA