Trichloroethylene, >99.5%, Electronic grade, Alfa Aesar™

Manufacturer: Alfa Aesar 41963M6

Quantity 4 x 4L

 View more versions of this product

Catalog No. AA41963M6

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 79-01-6
    Physical Form Liquid
    Assay Percent Range >99.5%
    Molecular Formula ClCH=CCl2
    Formula Weight 131.39
    MDL Number MFCD00000838
    Chemical Name or Material Trichloroethylene
    Grade Electronic Grade

    Specifications

    Formula Weight 131.39
    Quantity 4 x 4L
    Color Colorless
    DOT Information DOT Class: 6.1, Packaging Group: II
    EINECS Number 201-167-4
    Physical Form Liquid
    Assay Percent Range >99.5%
    Ignition Point 410°C (770°F)
    Molecular Formula ClCH=CCl2
    MDL Number MFCD00000838
    CAS 79-01-6
    Health Hazard 1 H350-H341-H315-H319-H336-H412: May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Harmful to aquatic life with long lasting effects.
    Odor Chloroform-like
    Appearance Colorless
    Vapor Pressure 77 hPa (58mm Hg) at 20°C
    Chemical Name or Material Trichloroethylene
    Grade Electronic Grade

    H350-H341-H315-H319-H336-H412: May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Harmful to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit