Learn More
Trichloroethylene, Electronic Grade, 99.5+%, Thermo Scientific Chemicals
Description
Trichloroethylene is used as solvent for paints, dyes, waxes, oils, fats and resins, as an additive in polymerization reactions, as a reactant to prepare various halohydrocarbons (1,1,1,2-tetrafluoroethane) and other chemicals, as a coolant for cutting tantalum and acts as good heat transfer medium. It is used in fire extinguisher fluids and in the recovery of oils and greases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 85°C |
Color | Colorless |
Density | 1.46 g/cm3 at 20°C |
Boiling Point | 87°C |
Flash Point | None |
Ignition Point | 410°C (770°F) |
Odor | Chloroform-like |
Assay Percent Range | >99.5% |
Refractive Index | 1.476 |
Quantity | 1 L |
Show More |
Safety and Handling
Hazard Category | H315-H317-H319-H336-H341-H350-H373 |
Precautionary Statement | P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c |
DOTInformation | DOT Class: 6.1; Packaging Group: III |
EINECSNumber | 201-167-4 |
RTECSNumber | KX4550000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.