Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroethylene, anhydrous, >99%, Thermo Scientific™

Catalog Number AA41894K2
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
79-01-6
C2HCl3
131.38
MFCD00000838
XSTXAVWGXDQKEL-UHFFFAOYSA-N
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
6575
CHEBI:16602
1,1,2-trichloroethene
C(=C(Cl)Cl)Cl
79-01-6
C2HCl3
131.38
MFCD00000838
XSTXAVWGXDQKEL-UHFFFAOYSA-N
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
6575
CHEBI:16602
1,1,2-trichloroethene
C(=C(Cl)Cl)Cl

Contains <0.01% water

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.46g/cm3 at 20°C
Ignition Point 410°C (770°F)
Odor Chloroform-like
Assay Percent Range >99%
Quantity 1 L
Vapor Pressure 77 hPa (58mm Hg) at 20°C
Formula Weight 131.39
Physical Form Liquid
Chemical Name or Material Trichloroethylene
Hazard Category H350-H341-H315-H319-H336-H412: May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Harmful to aquatic life with long lasting effects.
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 201-167-4
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit