Alfa Aesar™ Trichloroethylene, Semiconductor Grade, 99+%

Manufacturer: Alfa Aesar™ 39744K7

Quantity 4L

 View more versions of this product

Catalog No. AA39744K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 79-01-6
    Formula Weight 131.39
    MDL Number MFCD00000838
    UN Number UN1710
    Beilstein 1736782
    Merck Index 14,9639
    Chemical Name or Material Trichloroethylene
    Grade Semiconductor Grade

    Specifications

    Formula Weight 131.39
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1710
    Physical Form Liquid
    MDL Number MFCD00000838
    Odor Chloroform-like
    Refractive Index 1.476
    Melting Point −85°C
    Quantity 4L
    Density 1.46g/mL
    Beilstein 1736782
    Merck Index 14,9639
    Health Hazard 1 Danger
    Boiling Point 87°C
    TSCA TSCA
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE
    RTECS Number KX4550000
    CAS 79-01-6
    Health Hazard 2 GHS H Statement
    H350-H341-H315-H319-H336
    May cause cancer.
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    Chemical Name or Material Trichloroethylene
    Assay ≥99%
    Grade Semiconductor Grade

    GHS H Statement
    H350-H341-H315-H319-H336
    May cause cancer.
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit