Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trichloroethylene, Semiconductor Grade, 99+%

Manufacturer:  Alfa Aesar™ 39744K2

Quantity 1L

 View more versions of this product

Catalog No. AA39744K2


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

79-01-6
MFCD00000838
1736782
Trichloroethylene
131.39
UN1710
14,9639
Semiconductor Grade

Specifications

1L
1.46g/mL
131.39
UN1710
14,9639
1.476
−85°C
≥99%
79-01-6
Liquid
MFCD00000838
1736782
Chloroform-like
87°C
Trichloroethylene
Semiconductor Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H350-H341-H315-H319-H336
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE

RTECSNumber : KX4550000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit