Trichloroethylene, Spectrophotometric Grade, >99.5%, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Residue after evaporation 3ppm, H2O 0.02%

Manufacturer: Alfa Aesar 43487M6

Quantity 4 x 4L

 View more versions of this product

Catalog No. AA43487M6

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 79-01-6
    Physical Form Liquid
    Assay Percent Range >99.5%
    Molecular Formula ClCH=CCl2
    Formula Weight 131.39
    MDL Number MFCD00000838
    Chemical Name or Material Trichloroethylene
    Grade Spectrophotometric grade

    Specifications

    Physical Form Liquid
    Formula Weight 131.39
    Color Clear colorless
    Quantity 4 x 4L
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 201-167-4
    Ignition Point 410°C (770°F)
    Odor Chloroform-like
    Molecular Formula ClCH=CCl2
    MDL Number MFCD00000838
    CAS 79-01-6
    Vapor Pressure 58mm Hg at 20°C
    Health Hazard 1 H350-H341-H315-H319-H336-H412: May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Harmful to aquatic life with long lasting effects.
    Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
    Assay Percent Range >99.5%
    Chemical Name or Material Trichloroethylene
    Grade Spectrophotometric grade

    Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H350-H341-H315-H319-H336-H412: May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Harmful to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit