Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroethylene, Spectrophotometric Grade, >99.5%, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Residue after evaporation 3ppm, H2O 0.02%

Manufacturer:  Alfa Aesar 43487K7

Quantity 4L

 View more versions of this product

Catalog No. AA43487K7


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

79-01-6
>99.5%
131.39
Trichloroethylene
Liquid
ClCH=CCl2
MFCD00000838
Spectrophotometric grade

Specifications

79-01-6
>99.5%
131.39
410°C (770°F)
58mm Hg at 20°C
4L
Spectrophotometric grade
Liquid
ClCH=CCl2
MFCD00000838
Chloroform-like
Clear colorless
Trichloroethylene

Safety and Handling

SDS & Safety Documents

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.

H350-H341-H315-H319-H336-H412: May cause cancer. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause drowsiness or dizziness. Harmful to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 201-167-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit