Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroethylene, Stabilized, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
79-01-6
C2HCl3
131.38
XSTXAVWGXDQKEL-UHFFFAOYSA-N
trichloroethylene,trichloroethene,ethene, trichloro,trilene,trichlorethylene,ethinyl trichloride,ethylene trichloride,acetylene trichloride,chlorilen,narcogen
6575
CHEBI:16602
1,1,2-trichloroethene
C(=C(Cl)Cl)Cl
79-01-6
C2HCl3
131.38
XSTXAVWGXDQKEL-UHFFFAOYSA-N
trichloroethylene,trichloroethene,ethene, trichloro,trilene,trichlorethylene,ethinyl trichloride,ethylene trichloride,acetylene trichloride,chlorilen,narcogen
6575
CHEBI:16602
1,1,2-trichloroethene
C(=C(Cl)Cl)Cl

High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Packaging Aluminum Bottle
Specific Gravity 1L = 1.47kg
Formula Weight 131.39
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Trichloroethylene, Stabilized