Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trichloroisocyanuric acid, 90+%

Click to view available options
Quantity:
100g
500g
2500g
87-90-1
C3Cl3N3O3
232.401
MFCD00006553
YRIZYWQGELRKNT-UHFFFAOYSA-N
trichloroisocyanuric acid,symclosene,trichlorocyanuric acid,symclosen,isocyanuric chloride,fichlor 91,trichloro-s-triazinetrione,trichloroisocyanic acid,fi clor 91,chloreal
6909
CHEBI:33015
1,3,5-trichloro-1,3,5-triazinane-2,4,6-trione
C1(=O)N(C(=O)N(C(=O)N1Cl)Cl)Cl
87-90-1
C3Cl3N3O3
232.401
MFCD00006553
YRIZYWQGELRKNT-UHFFFAOYSA-N
trichloroisocyanuric acid,symclosene,trichlorocyanuric acid,symclosen,isocyanuric chloride,fichlor 91,trichloro-s-triazinetrione,trichloroisocyanic acid,fi clor 91,chloreal
6909
CHEBI:33015
1,3,5-trichloro-1,3,5-triazinane-2,4,6-trione
C1(=O)N(C(=O)N(C(=O)N1Cl)Cl)Cl
Applications
Trichloroisocyanuric acid was used as a disinfection reagent in the transformation of benzophenone-3 in chlorinated water. It was also used in the synthesis of 2-halomethylene-3-oxoketoximes. It is used as a bleaching agent in the textile industry, a reagent in organic synthesis.

Solubility
Soluble in chlorocarbon, acetonitrile, acetone, water (12 g/L).

Notes
Moisture sensitive. Store away from reducing agents, acids, bases, flammable substances, organic materials, light, water/ moisture.
pH 3
Odor Pungent
Flash Point >250°C (482°F)
Melting Point ~245°C (decomposition)
Quantity 500g
UN Number UN2468
Beilstein 202022
Merck Index 14,9641
Formula Weight 232.41
Percent Purity ≥90%
Chemical Name or Material Trichloroisocyanuric acid
Show More Show Less

GHS H Statement
H272-H302-H319-H335
May intensify fire; oxidizer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: TRICHLOROISOCYANURIC ACID, DRY

RTECSNumber : XZ1925000

TSCA : TSCA