Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichlorovinylsilane, 96%, ACROS Organics™

Click to view available options
Quantity:
10mL
Packaging:
Glass bottle
75-94-5
C2H3Cl3Si
161.49
MFCD00000482
GQIUQDDJKHLHTB-UHFFFAOYSA-N
trichlorovinylsilane,vinyltrichlorosilane,trichloro vinyl silane,vtcs,silane, trichloroethenyl,silane, trichlorovinyl,trichlorovinylsilicon,vinyl trichlorosilane,trichlorovinyl silicane,vinylsilicon trichloride
6413
trichloro(ethenyl)silane
C=C[Si](Cl)(Cl)Cl
75-94-5
C2H3Cl3Si
161.49
MFCD00000482
GQIUQDDJKHLHTB-UHFFFAOYSA-N
trichlorovinylsilane,vinyltrichlorosilane,trichloro vinyl silane,vtcs,silane, trichloroethenyl,silane, trichlorovinyl,trichlorovinylsilicon,vinyl trichlorosilane,trichlorovinyl silicane,vinylsilicon trichloride
6413
trichloro(ethenyl)silane
C=C[Si](Cl)(Cl)Cl
Viscosity 0.6 mPa.s (20°C)
Refractive Index 1.4350 to 1.4370
Specific Gravity 1.27
Infrared Spectrum Authentic
Melting Point -95.0°C
Quantity 10mL
Boiling Point 90.0°C
Packaging Glass bottle
Flash Point 10°C
Density 1.2700g/mL
Color Colorless to Brown
Assay Percent Range 96%
Linear Formula H2C=CHSiCl3
Fieser 05,749
Solubility Information Solubility in water: reacts
Formula Weight 161.49
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Trichlorovinylsilane, 96%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-917-8

RTECSNumber : VV6125000

TSCA : TSCA