Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tricine, ≥98.5% (Dry Basis), Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Plastic bottle
5704-04-1
C6H13NO5
179.17
MFCD00004277
SEQKRHFRPICQDD-UHFFFAOYSA-N
N-Tris(Hydroxymethyl)methylglycine
79784
CHEBI:39063
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C(C(=O)O)NC(CO)(CO)CO
5704-04-1
C6H13NO5
179.17
MFCD00004277
SEQKRHFRPICQDD-UHFFFAOYSA-N
N-Tris(Hydroxymethyl)methylglycine
79784
CHEBI:39063
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C(C(=O)O)NC(CO)(CO)CO
Linear Formula (CH2OH)3CNHCH2COOH
Formula Weight 179.17
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Packaging Plastic bottle
Melting Point ~183°C (decomposition)
Quantity 100g
Chemical Name or Material Tricine
Assay Dry basis