Learn More
Research Products International CorpSupplier Diversity Partner Tricine, Ultra Pure [N-[Tris (Hydroxymethyl) Methyl] Glycine], 1 Kilogram DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner T240001000.0
pKa @ 25?C: 8.1-8.2 Melting Range: 182-188?C
Catalog No. 50-489-072