Learn More
Cytiva Tricorn Columns PROMO
Tricorn columns are compatible with high flow rates and are designed for high-performance media such as Sepharose, Superdex, and SOURCE.
$640.80 - $667.80
Specifications
For Use With (Application) High resolution liquid chromatography of biomolecules performed at medium to high pressures
Includes Glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock and filter holder
Material Borosilicate glass
Packaging Quantity 1 Ea.
Support Type Polypropylene filter
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity & Availability  
45-200-39LR1
View Documents
Cytiva
29648093
5 mm
Each for $640.80
Add to cart
45-200-45LR1
View Documents
Cytiva
29648094
10 mm
Each for $667.80
Add to cart
Description
Description
  • Tricorn empty columns are designed for high performance separations on small and medium scale
  • The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
  • Broad range of column dimensions suitable for many different purification needs.
  • Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
  • Separate packing connector allows simple column packing for a broad range of media.
  • Locking ring prevents accidental compression of media bed.
Specifications
Specifications
High resolution liquid chromatography of biomolecules performed at medium to high pressures
Borosilicate glass
Polypropylene filter
Glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock and filter holder
1 Ea.
Tricorn Columns (Empty)