Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Tricorn™ Superdex™ Prepacked Gel Filtration Columns

Use for laboratory-scale separations and intermediate purification based on different sizes of proteins, peptides, and other biomolecules. Cytiva Tricorn™ Superdex™ Prepacked Gel Filtration Columns are perfect for high-resolution applications.

Specifications

Buffer 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol, 70% Ethanol, 1M NaOH
Quantity Each
For Use With (Application) High-resolution, semipreparative and analytical separations of biomolecules
Includes Valco fittings for AKTA™ design systems and are supplied with M6 connecto rs for FPLC System
Material Borosilicate Glass
View More Specs

Includes: Valco fittings for AKTAdesign systems and are supplied with M6 connectors for FPLC System.

Products 2
Catalog Number Mfr. No. Product Type Flow Rate Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Flow Rate Height (English) Price Quantity & Availability  
45-002-903
View Documents Promotion Details
Cytiva
17517401
Superdex™ 75 prepacked column 0.5mL/min. 11.81 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
45-002-905
View Documents Promotion Details
Cytiva
17517601
Superdex™ Peptide prepacked column 0.15 to 0.6mL/min. 11.81 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Fast gel filtration involving separation based on three molecular weight ranges from 100 to 600000 M
  • Spherical composite of cross-linked agarose and dextran matrix
  • Comes with two different bed heights of 150 and 300mm allowing greater flexibility to suit different analytical needs
  • Available with three different media: Superdex Peptide, Superdex 75, and Superdex 200

Superdex Peptide:

  • Bed dimension: 10 x 300mm
  • Bed volume: 24mL
  • Flow velocity: 0.15 to 0.6mL/min.
  • Pressure max.: 18 bar (261psi)
  • Separation range: 100 to 7000 (peptides)
  • pH Stability working range: 1 to 14
  • Column I.D.: 10mm
  • Bed height with one adaptor:     279 to 309mm
  • Bed height with two adaptors: 245 to 306mm
Superdex 200:
  • Bed dimension: 5 x 150mm and 10 x 300mm
  • Bed volume: 3 and 24mL
  • Flow velocity: 0.15 to 0.6mL/min. and 0.25 to 0.75mL/min.
  • Pressure max.: 15 bar (217psi)
  • Separation range: 10,000 to 600,000 (globular peptides)
  • pH Stability working range: 3 to 12
  • Column I.D.: 5 and 10mm
  • Bed height with one adaptor:     135 to 159mm (3mL column)
  • Bed height with two adaptors: 108 to 156mm (3mL column)
Superdex 75:
  • Bed dimension: 5 x 150mm and 10 x 300mm
  • Bed volume: 3 and 24mL
  • Flow velocity: 0.15 to 0.6mL/min.
  • Pressure max.: 18 bar (261psi)
  • Separation range: 3000 up to 70,000 (globular proteins)
  • pH Stability working range: 3 to 12
  • Column I.D.: 5 and 10 mm
  • Bed height with one adaptor: 108 to 156mm (3mL column)
  • Bed height with two adaptors: 135 to 159mm (3mL column)

Specifications

Specifications

1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol, 70% Ethanol, 1M NaOH
High-resolution, semipreparative and analytical separations of biomolecules
Borosilicate Glass
13μm
Spherical composite of cross-linked Agarose and Dextran
Each
Valco fittings for AKTA™ design systems and are supplied with M6 connecto rs for FPLC System
18 bar
4° to 30°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit