Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tricyclohexylphosphine, technical grade, Thermo Scientific Chemicals

Catalog Number AA4485403
Click to view available options
Quantity:
1 g
5 g
2622-14-2
C18H33P
280.436
MFCD00003853
WLPUWLXVBWGYMZ-UHFFFAOYSA-N
tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
75806
tricyclohexylphosphane
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3
2622-14-2
C18H33P
280.436
MFCD00003853
WLPUWLXVBWGYMZ-UHFFFAOYSA-N
tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
75806
tricyclohexylphosphane
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 76°C to 78°C
Color White
Odor Unpleasant
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 280.42
Grade Technical
Physical Form Solid
Chemical Name or Material Tricyclohexylphosphine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-069-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit