Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tricyclohexylphosphonium tetrafluoroborate, 99%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
58656-04-5
C18H34BF4P
368.24
MFCD03840580
MYSMMEUXKHJYKH-UHFFFAOYSA-O
tricyclohexylphosphonium tetrafluoroborate,tricyclohexylphosphine tetrafluoroborate,tricyclohexylphosphine tetrafluroborate,tricyclohexylphosphoniumtetrafluoroborate,pcy3.hbf4,ksc269e4n,tricyclohexylphosphine tetrofluroborate,tricyclohexyl phosphonium tetrafluoroborate,tetrafluoroboranuide tricyclohexylphosphanium,tricyclohexyl-phosphonium tetrafluoro borate
11710396
tricyclohexylphosphanium;tetrafluoroborate
[B-](F)(F)(F)F.C1CCC(CC1)[PH+](C2CCCCC2)C3CCCCC3
58656-04-5
C18H34BF4P
368.24
MFCD03840580
MYSMMEUXKHJYKH-UHFFFAOYSA-O
tricyclohexylphosphonium tetrafluoroborate,tricyclohexylphosphine tetrafluoroborate,tricyclohexylphosphine tetrafluroborate,tricyclohexylphosphoniumtetrafluoroborate,pcy3.hbf4,ksc269e4n,tricyclohexylphosphine tetrofluroborate,tricyclohexyl phosphonium tetrafluoroborate,tetrafluoroboranuide tricyclohexylphosphanium,tricyclohexyl-phosphonium tetrafluoro borate
11710396
tricyclohexylphosphanium;tetrafluoroborate
[B-](F)(F)(F)F.C1CCC(CC1)[PH+](C2CCCCC2)C3CCCCC3
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color White
Melting Point 165.0°C to 169.0°C
Elemental Analysis 98.5% min.
Assay Percent Range 99%
Solubility Information Solubility in water: soluble
Formula Weight 368.24
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Tricyclohexylphosphonium tetrafluoroborate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.