Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tricyclohexyltin chloride, 97%, Thermo Scientific Chemicals

Catalog Number AAH5549314
Click to view available options
Quantity:
1 g
5 g
25 g
3091-32-5
C18H33ClSn
403.622
MFCD00003817
OJVZYIKTLISAIH-UHFFFAOYSA-M
tricyclohexyltin chloride, stannane, chlorotricyclohexyl, chlorotricyclohexyltin, chloro tricyclohexyl stannane, tin, chlorotricyclohexyl, tricyclohexyltinchloride, acmc-1ck6z, tricyclohexylchlorotin
76539
chloro(tricyclohexyl)stannane
C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl
3091-32-5
C18H33ClSn
403.622
MFCD00003817
OJVZYIKTLISAIH-UHFFFAOYSA-M
tricyclohexyltin chloride, stannane, chlorotricyclohexyl, chlorotricyclohexyltin, chloro tricyclohexyl stannane, tin, chlorotricyclohexyl, tricyclohexyltinchloride, acmc-1ck6z, tricyclohexylchlorotin
76539
chloro(tricyclohexyl)stannane
C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl

Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 123°C to 125°C
Linear Formula (C6H11)3SnCl
Quantity 25 g
UN Number UN3146
Solubility Information Soluble in ethanol,ether,toluene and chloroform. Insoluble in water.
Formula Weight 403.6
Percent Purity 97%
Chemical Name or Material Tricyclohexyltin chloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 221-437-5
RTECSNumber WH6830000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit