Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridecanal, 96%, stab., Alfa Aesar™

Click to view available options
Quantity:
5g
25g
10486-19-8
C13H26O
198.35
MFCD00007018
BGEHHAVMRVXCGR-UHFFFAOYSA-N
tridecyl aldehyde,tridecanaldehyde,1-tridecanal,n-tridecylaldehyde,n-tridecanal,tridecane aldehyde,tridecanealdehyde,aldehyde c-13,technical,tridecanal
25311
tridecanal
CCCCCCCCCCCCC=O
10486-19-8
C13H26O
198.35
MFCD00007018
BGEHHAVMRVXCGR-UHFFFAOYSA-N
tridecyl aldehyde,tridecanaldehyde,1-tridecanal,n-tridecylaldehyde,n-tridecanal,tridecane aldehyde,tridecanealdehyde,aldehyde c-13,technical,tridecanal
25311
tridecanal
CCCCCCCCCCCCC=O
Sensitivity Air sensitive
Refractive Index 1.4384
Density 0.83
Melting Point 14°C to 15°C
Flash Point 132°C (269°F)
Boiling Point 280°C
Odor Characteristic
Quantity 25g
Assay Percent Range 0.94
Linear Formula CH3(CH2)11CHO
Formula Weight 198.35
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Tridecanal, stabilized
Show More Show Less

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 234-004-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen