Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethanolamine 97%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
102-71-6
C6H15NO3
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine,trolamine,2,2',2-nitrilotriethanol,sterolamide,daltogen,tris 2-hydroxyethyl amine,triethylolamine,nitrilotriethanol,trihydroxytriethylamine,thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO
102-71-6
C6H15NO3
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine,trolamine,2,2',2-nitrilotriethanol,sterolamide,daltogen,tris 2-hydroxyethyl amine,triethylolamine,nitrilotriethanol,trihydroxytriethylamine,thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO
Refractive Index 1.4840 to 1.4860
Flash Point 190°C
Specific Gravity 1.1245
Density 1.1245g/mL
Quantity 1L
Vapor Pressure <0.01mmHg at 20°C
Melting Point 21.0°C
Boiling Point 360.0°C
Infrared Spectrum Authentic
Color Undesignated
Decomposition Information >325°C
Assay Percent Range 97%
Linear Formula (HOCH2CH2)3N
Fieser 01,1196
Merck Index 15, 9830
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with alcohol, methanol and acetone, soluble in chloroform, slichtly soluble in benzene and ether, soluble 4.2% in benzene; 1.6% in ether; 0.4% in, carbon tetrachloride and < 0.1% in n-hepta
Formula Weight 149.19
Physical Form Oily Liquid
Percent Purity 97%
Chemical Name or Material Triethanolamine, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 203-049-8

RTECSNumber : KL9275000

TSCA : TSCA