Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triethanolamine hydrochloride, 99+%

Click to view available options
Quantity:
250g
1000g
5000g
637-39-8
C6H16ClNO3
185.648
MFCD00012596
HHLJUSLZGFYWKW-UHFFFAOYSA-N
triethanolamine hydrochloride,tea-hydrochloride,triethanolammonium chloride,2,2',2-nitrilotriethanol hydrochloride,ethanol, 2,2',2-nitrilotris-, hydrochloride,2,2',2-nitrilotrisethanol hydrochloride,unii-r297uj9qdy,tris 2-hydroxyethyl ammonium chloride,tris 2-hydroxyethyl amine hydrochloride,r297uj9qdy
101814
2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride
C(CO)N(CCO)CCO.Cl
637-39-8
C6H16ClNO3
185.648
MFCD00012596
HHLJUSLZGFYWKW-UHFFFAOYSA-N
triethanolamine hydrochloride,tea-hydrochloride,triethanolammonium chloride,2,2',2-nitrilotriethanol hydrochloride,ethanol, 2,2',2-nitrilotris-, hydrochloride,2,2',2-nitrilotrisethanol hydrochloride,unii-r297uj9qdy,tris 2-hydroxyethyl ammonium chloride,tris 2-hydroxyethyl amine hydrochloride,r297uj9qdy
101814
2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride
C(CO)N(CCO)CCO.Cl
Applications
Protein purification reagent Triethanolamine hydrochloride serves as a protein purification reagent, an emulsifier and a surfactant. It is used in the study of chemical cross linking of chloroplast coupling factor and acts as a medium for assaying adenosine diphosphate (ADP). It acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Quantity 250g
Sensitivity Hygroscopic
Melting Point 177°C to 179°C
Linear Formula (HOCH2CH2)3N·HCl
Beilstein 3909940
Merck Index 14,9665
Solubility Information Soluble in water.
Formula Weight 185.65
Percent Purity ≥99%
Chemical Name or Material Triethanolamine hydrochloride

EINECSNumber : 211-284-2

RTECSNumber : KL9346500

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen