Alfa Aesar™ Triethoxysilane, 96%

Manufacturer: Alfa Aesar™ B2206309

Quantity 10g

 View more versions of this product

Catalog No. AAB2206309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 998-30-1
    Molecular Formula C6H16O3Si
    Linear Formula (CH3CH2O)3SiH
    Formula Weight 164.28
    MDL Number MFCD00009061
    UN Number UN3384
    Beilstein 1738989
    Chemical Name or Material Triethoxysilane

    Specifications

    Formula Weight 164.28
    Molecular Formula C6H16O3Si
    UN Number UN3384
    Beilstein 1738989
    Quantity 10g
    Flash Point 26°C (78°F)
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
    TSCA TSCA
    Linear Formula (CH3CH2O)3SiH
    MDL Number MFCD00009061
    CAS 998-30-1
    Boiling Point 134° to 135°C
    Melting Point −170°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H330-H314-H318-H226
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    RTECS Number VV6682000
    Chemical Name or Material Triethoxysilane
    Assay 96%

    Description

    Applications
    Triethoxysilane is raw material for synthesis of silane addition agent and silicon oil. It is used as a reducing agent.

    Solubility
    Insoluble in water. Soluble in organic solvents.

    Notes
    Moisture sensitive. Store away from strong oxidizing agents, strong acids, bases, water/ moisture. Store under inert gas.

    GHS H Statement
    H330-H314-H318-H226
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.

    GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit