Alfa Aesar™ Triethyl 1,3,5-benzenetricarboxylate, 97%

Manufacturer: Alfa Aesar™ B2339222

Quantity 100g

 View more versions of this product

Catalog No. AAB2339222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4105-92-4
    Molecular Formula C15H18O6
    Formula Weight 294.31
    MDL Number MFCD00236389
    Synonym 1,3,5-Benzenetricarboxylic acid triethyl ester; Triethyl trimesate
    Chemical Name or Material Triethyl 1,3,5-benzenetricarboxylate

    Specifications

    Formula Weight 294.31
    Molecular Formula C15H18O6
    MDL Number MFCD00236389
    CAS 4105-92-4
    Synonym 1,3,5-Benzenetricarboxylic acid triethyl ester; Triethyl trimesate
    Melting Point 132° to 135°C
    Quantity 100g
    Chemical Name or Material Triethyl 1,3,5-benzenetricarboxylate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit