Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl 4-phosphonobutyrate, 93%, Acros Organics

Click to view available options
Quantity:
25g
2327-69-7
C10H21O5P
252.247
MFCD00015168
FYESTJSRASJZOY-UHFFFAOYSA-N
triethyl 4-phosphonobutyrate,ethyl 4-diethoxyphosphoryl butanoate,triethyl4-phosphonobutyrate,butanoic acid, 4-diethoxyphosphinyl-, ethyl ester,triethyl-4-phosphono butyrate,butyric acid, 4-diethylphosphono-, ethyl ester,triethyl 4-phosphonobutanoate,4-phosphonobutanoic acid triethyl ester,4-diethoxyphosphinyl butyric acid ethyl ester,butanoic acid,4-diethoxyphosphinyl-,ethyl ester
281201
ethyl 4-diethoxyphosphorylbutanoate
CCOC(=O)CCCP(=O)(OCC)OCC
2327-69-7
C10H21O5P
252.247
MFCD00015168
FYESTJSRASJZOY-UHFFFAOYSA-N
triethyl 4-phosphonobutyrate,ethyl 4-diethoxyphosphoryl butanoate,triethyl4-phosphonobutyrate,butanoic acid, 4-diethoxyphosphinyl-, ethyl ester,triethyl-4-phosphono butyrate,butyric acid, 4-diethylphosphono-, ethyl ester,triethyl 4-phosphonobutanoate,4-phosphonobutanoic acid triethyl ester,4-diethoxyphosphinyl butyric acid ethyl ester,butanoic acid,4-diethoxyphosphinyl-,ethyl ester
281201
ethyl 4-diethoxyphosphorylbutanoate
CCOC(=O)CCCP(=O)(OCC)OCC
for C-C Bond Formation (Olefination)
Refractive Index 1.4360 to 1.4400 (20°C, 589nm)
Color Yellow
Quantity 25g
Infrared Spectrum Authentic
Assay Percent Range 93%
Linear Formula (O)P(CH3CHO)(CH)3C(O)OCHCH3
Formula Weight 252.25
Physical Form Liquid
Percent Purity ≥91%
Chemical Name or Material Triethyl 4-phosphonobutyrate