Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl 4-phosphonocrotonate, 92%, predominantly trans, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
10236-14-3
C10H19O5P
250.231
MFCD00009192
LXLODBXSCRTXFG-BQYQJAHWSA-N
triethyl 4-phosphonocrotonate,e-ethyl 4-diethoxyphosphoryl but-2-enoate,ethyl 4-diethoxyphosphoryl but-2-enoate,triethyl .gamma.-phosphonocrotonate,ethyl 4-diethylphosphono crotonate,ethyl 2e-4-diethoxyphosphoryl but-2-enoate,ethyl e-4-diethoxyphosphorylbut-2-enoate,ethyl 4-diethylphosphonocrotonate,ethyl trans-4-diethylphosphono crotonate,4-diethoxyphosphinyl crotonic acid ethyl ester
5365783
ethyl (E)-4-diethoxyphosphorylbut-2-enoate
CCOC(=O)C=CCP(=O)(OCC)OCC
10236-14-3
C10H19O5P
250.231
MFCD00009192
LXLODBXSCRTXFG-BQYQJAHWSA-N
triethyl 4-phosphonocrotonate,e-ethyl 4-diethoxyphosphoryl but-2-enoate,ethyl 4-diethoxyphosphoryl but-2-enoate,triethyl .gamma.-phosphonocrotonate,ethyl 4-diethylphosphono crotonate,ethyl 2e-4-diethoxyphosphoryl but-2-enoate,ethyl e-4-diethoxyphosphorylbut-2-enoate,ethyl 4-diethylphosphonocrotonate,ethyl trans-4-diethylphosphono crotonate,4-diethoxyphosphinyl crotonic acid ethyl ester
5365783
ethyl (E)-4-diethoxyphosphorylbut-2-enoate
CCOC(=O)C=CCP(=O)(OCC)OCC
For C-C Bond Formation (Olefination)
Color Yellow
Refractive Index 1.4525 to 1.4555 (20°C, 589nm)
Quantity 1g
Infrared Spectrum Authentic
Density 1.1200g/mL
Assay Percent Range 92%
Linear Formula (C2H5O)2P(O)CH2CH=CHCO2C2H5
Formula Weight 250.23
Physical Form Liquid
Percent Purity ≥90%
Chemical Name or Material Triethyl 4-phosphonocrotonate, 92%
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning