Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl 4-phosphonocrotonate, 92%, predominantly trans, Thermo Scientific Chemicals

Catalog Number AC152120500
Click to view available options
Quantity:
10 g
50 g
10236-14-3
C10H19O5P
250.23
MFCD00009192
LXLODBXSCRTXFG-BQYQJAHWSA-N
triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
5365783
CCOC(=O)\C=C\CP(=O)(OCC)OCC
10236-14-3
C10H19O5P
250.23
MFCD00009192
LXLODBXSCRTXFG-BQYQJAHWSA-N
triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
5365783
CCOC(=O)\C=C\CP(=O)(OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.1200g/mL
Boiling Point 133.0°C to 135.0°C (0.4 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 92%
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH2CH=CHCO2C2H5
Refractive Index 1.4525 to 1.4555
Quantity 50 g
Specific Gravity 1.12
IUPAC Name ethyl (E)-4-diethoxyphosphorylbut-2-enoate
Formula Weight 250.23
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material Triethyl 4-phosphonocrotonate, 92%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit