Alfa Aesar™ Triethyl 4-phosphonocrotonate, cis + trans, 94%

Manufacturer: Alfa Aesar™ A1099530

Quantity 250g

 View more versions of this product

Catalog No. AAA1099530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10236-14-3
    Molecular Formula C10H19O5P
    Formula Weight 250.23
    MDL Number MFCD00009192
    Synonym 4-Phosphonocrotonic acid triethyl ester; Triethyl trans-4-phosphono-2-butenoate
    Chemical Name or Material Triethyl 4-phosphonocrotonate, cis + trans

    Specifications

    Formula Weight 250.23
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C10H19O5P
    CAS 10236-14-3
    Synonym 4-Phosphonocrotonic acid triethyl ester; Triethyl trans-4-phosphono-2-butenoate
    Boiling Point 133° to 135°C (0.4mmHg)
    MDL Number MFCD00009192
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point >110°C (230°F)
    Quantity 250g
    Health Hazard 1 Warning
    Chemical Name or Material Triethyl 4-phosphonocrotonate, cis + trans
    Assay 94%

    Description

    Applications
    Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit