Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl Borate 97%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
150-46-9
C6H15BO3
145.99
MFCD00009073
AJSTXXYNEIHPMD-UHFFFAOYSA-N
triethoxyborane,boron ethoxide,boron triethoxide,triethoxyboron,boric acid, triethyl ester,boric acid triethyl ester,borane, triethoxy,triethylborate,ethyl borate eto 3b,boric acid h3bo3 , triethyl ester
9009
CHEBI:38916
triethyl borate
B(OCC)(OCC)OCC
150-46-9
C6H15BO3
145.99
MFCD00009073
AJSTXXYNEIHPMD-UHFFFAOYSA-N
triethoxyborane,boron ethoxide,boron triethoxide,triethoxyboron,boric acid, triethyl ester,boric acid triethyl ester,borane, triethoxy,triethylborate,ethyl borate eto 3b,boric acid h3bo3 , triethyl ester
9009
CHEBI:38916
triethyl borate
B(OCC)(OCC)OCC
Specific Gravity 0.858
Refractive Index 1.3720 to 1.3740
Melting Point -84.5°C
Flash Point 11°C
Quantity 1L
Density 0.8580g/mL
Boiling Point 117.0°C to 118.0°C
Infrared Spectrum Authentic
Color Colorless
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula (C2H5O)3B
Beilstein 01, 335
Solubility Information Solubility in water: decomposes
Formula Weight 145.99
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Triethyl borate, 97%
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 205-760-9

RTECSNumber : ED5075000

TSCA : TSCA