Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triethyl Citrate, FCC, 99-100.5%, Spectrum™ Chemical
Click to view available options
Quantity:
500 mL
4 L
5 gal.
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Pail
Description
Spectrum™ Chemical Triethyl Citrate, FCC is a colorless, odorless liquid used as a food additive (E number E1505) to stabilize foams, especially as whipping aid for egg whites. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 1 |
Packaging | Amber Glass Bottle |
Refractive Index | 1.439-1.443 |
Quantity | 4 L |
Percent Purity | 99.0 to 100.5% |
Grade | FCC |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction