Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl orthobenzoate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1663-61-2
C13H20O3
224.3
MFCD00009222
BQFPCTXLBRVFJL-UHFFFAOYSA-N
triethoxymethyl benzene,triethyl orthobenzoate,ethyl orthobenzoate,benzene, triethoxymethyl,triethylorthobenzoate,orthobenzoic acid, triethyl ester,orthobenzoic acid triethyl ester,triethoxymethyl benzol,triethoxyphenylmethane
74268
triethoxymethylbenzene
CCOC(C1=CC=CC=C1)(OCC)OCC
1663-61-2
C13H20O3
224.3
MFCD00009222
BQFPCTXLBRVFJL-UHFFFAOYSA-N
triethoxymethyl benzene,triethyl orthobenzoate,ethyl orthobenzoate,benzene, triethoxymethyl,triethylorthobenzoate,orthobenzoic acid, triethyl ester,orthobenzoic acid triethyl ester,triethoxymethyl benzol,triethoxyphenylmethane
74268
triethoxymethylbenzene
CCOC(C1=CC=CC=C1)(OCC)OCC
Alkylation reagent
Flash Point 96°C
Boiling Point 239°C to 241°C
Specific Gravity 0.991
Density 0.9910g/mL
Refractive Index 1.4710 to 1.474
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
Assay Percent Range 96% min. (GC)
Linear Formula C6H5C(OCH2CH3)3
Solubility Information Solubility in water: hydrolysis
Formula Weight 224.3
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Triethyl orthobenzoate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-771-3