Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl orthoformate, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
2.5L
25L
Packaging:
Glass bottle
Metal Drum
122-51-0
C7H16O3
148.2
MFCD00009230
GKASDNZWUGIAMG-UHFFFAOYSA-N
triethyl orthoformate,triethoxymethane,ethyl orthoformate,aethon,ethone,diethoxymethoxy ethane,methane, triethoxy,ethyl formate ortho,orthoformic acid triethyl ester,orthomravencan ethylnaty
31214
diethoxymethoxyethane
CCOC(OCC)OCC
122-51-0
C7H16O3
148.2
MFCD00009230
GKASDNZWUGIAMG-UHFFFAOYSA-N
triethyl orthoformate,triethoxymethane,ethyl orthoformate,aethon,ethone,diethoxymethoxy ethane,methane, triethoxy,ethyl formate ortho,orthoformic acid triethyl ester,orthomravencan ethylnaty
31214
diethoxymethoxyethane
CCOC(OCC)OCC
Alkylation reagent
Specific Gravity 0.89
Refractive Index 1.3900 to 1.3920
Quantity 100mL
Packaging Glass bottle
Density 0.8900g/mL
Color Colorless
Boiling Point 146.0°C
Flash Point 35°C
Melting Point -61.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CH(OC2H5)3
Beilstein 02, 20
Fieser 01,1204; 02,430; 04,527; 05,690; 06,610; 07,385; 11,555; 13,327
Merck Index 15, 6980
Solubility Information Solubility in water: 1.35g/L (20°C). Other solubilities: decomposes in water,soluble in alcohols,acetone,ethers,chloroform,
Formula Weight 148.2
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Triethyl orthoformate
Show More Show Less

GHS H Statement:
Flammable liquid and vapour.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning