Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl orthopropionate, 98+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
115-80-0
C9H20O3
176.26
MFCD00009226
FGWYWKIOMUZSQF-UHFFFAOYSA-N
triethyl orthopropionate,ethyl orthopropionate,propane, 1,1,1-triethoxy,triethylorthopropionate,triethyl o-propionate,orthopropionic acid ethyl ester,orthopropionic acid, triethyl ester,unii-9uvm48bas9,9uvm48bas9,1,1,1-triethoxypropan
66995
1,1,1-triethoxypropane
CCC(OCC)(OCC)OCC
115-80-0
C9H20O3
176.26
MFCD00009226
FGWYWKIOMUZSQF-UHFFFAOYSA-N
triethyl orthopropionate,ethyl orthopropionate,propane, 1,1,1-triethoxy,triethylorthopropionate,triethyl o-propionate,orthopropionic acid ethyl ester,orthopropionic acid, triethyl ester,unii-9uvm48bas9,9uvm48bas9,1,1,1-triethoxypropan
66995
1,1,1-triethoxypropane
CCC(OCC)(OCC)OCC
Alkylation reagent
Color Colorless
Density 0.8860g/mL
Boiling Point 155.0°C to 160.0°C
Packaging Glass bottle
Quantity 100g
Infrared Spectrum Authentic
Flash Point 60°C
Specific Gravity 0.886
Refractive Index 1.4010 to 1.4030
Assay Percent Range 98% min. (GC)
Linear Formula C2H5C(OC2H5)3
Beilstein 02, 240
Solubility Information Solubility in water: decomposes
Formula Weight 176.26
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Triethyl orthopropionate, 98+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-108-0

TSCA : TSCA