Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triethyl phosphate, 99+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Description
Triethyl phosphate is used as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.072 g/mL |
Boiling Point | 215°C |
Flash Point | 115°C (240°F) |
Odor | Mild |
Refractive Index | 1.405 |
Quantity | 25 g |
Merck Index | 14,9674 |
Solubility Information | Soluble in water. |
Formula Weight | 182.16 |
Percent Purity | ≥99% |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | H302 |
missing translation for 'healthHazard2' | GHS H Statement H302 Harmful if swallowed. |
missing translation for 'healthHazard3' | P264b-P270-P301+P312-P330-P501c |
EINECSNumber | 201-114-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'